6-(Trifluoromethyl)oxindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IN-0082-5G
IN-0082-25G
COBBIN-0082-5GEA
0
GBP
COBBIN-0082-5G
COBBIN-0082-25G
6-(Trifluoromethyl)oxindole
6-(Trifluoromethyl)oxindole
Formula:
C₉H₆F₃NO |
MDL Number:
MFCD01075220 CAS Number: 1735-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...