5-(Trifluoromethyl)oxindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0076-5G IN-0076-1G
COBBIN-0076-5GEA 0 GBP
COBBIN-0076-5G COBBIN-0076-1G
5-(Trifluoromethyl)oxindole
5-(Trifluoromethyl)oxindole
Formula: C₉H₆F₃NO
MW: 201.15 g/mol
MDL Number: MFCD02179602
CAS Number: 71293-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR