5-Aminooxindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Aminoindolin-2-one, 5-Amino-1,3-dihydro-indol-2-one, 5-Amino-2-indolinone, 5-Aminooxindole

IN-0077-5G IN-0077-1G
COBBIN-0077-5GEA 0 GBP
COBBIN-0077-5G COBBIN-0077-1G
5-Aminooxindole
5-Aminooxindole
Formula: C₈H₈N₂O
MW: 148.16 g/mol
MDL Number: MFCD02179603
CAS Number: 20876-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR