5-Chlorooxindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chloro-2-oxindole

IN-0108-5G IN-0108-25G IN-0108-100G
COBBIN-0108-5GEA 0 GBP
COBBIN-0108-5G COBBIN-0108-25G COBBIN-0108-100G
5-Chlorooxindole
5-Chlorooxindole
Formula: C₈H₆ClNO
MW: 167.59 g/mol
MDL Number: MFCD00191927
CAS Number: 17630-75-0
EINECS: 412-200-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR