Ethyl piperazine-1-carboxylate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate
M-Clarity™ quality level = MQ100
E45600-100G
E45600-25G
SIALE45600-100GEA
99.8
GBP
SIALE45600-100G
SIALE45600-25G
Ethyl piperazine-1-carboxylate, Sigma-Aldrich®
Ethyl piperazine-1-carboxylate
Formula:
C₇H₁₄N₂O₂ MW: 158.2 g/mol |
MDL Number:
MFCD00005964 CAS Number: 120-43-4 EINECS: 204-395-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...