Geraniol

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

EHERC14010000EA 107 GBP
EHERC14010000
Geraniol
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR