Mycophenolic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

M-Clarity™ quality level = MQ300

M3536-250MG M3536-50MG
SIALM3536-250MGEA 570 GBP
SIALM3536-250MG SIALM3536-50MG
Mycophenolic acid, Sigma-Aldrich®
Mycophenolic acid
Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1
EINECS: 246-119-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR