3,5-Dibromoaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1207-5G
AN-1207-25G
AN-1207-1G
COBBAN-1207-5GEA
0
GBP
COBBAN-1207-5G
COBBAN-1207-25G
COBBAN-1207-1G
3,5-Dibromoaniline hydrochloride
3,5-Dibromoaniline hydrochloride
Formula:
C₆H₆Br₂ClN |
MDL Number:
MFCD09800855 CAS Number: 188349-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...