Dimethyl fluoromalonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dimethyl 2-fluoromalonate

001555-5G 001555-25G
FLUO001555-5GEA 0 GBP
FLUO001555-5G FLUO001555-25G
Dimethyl fluoromalonate
Dimethyl fluoromalonate
Formula: C₅H₇FO₄
MW: 150.11 g/mol
MDL Number: MFCD00054680
CAS Number: 344-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR