3,5-Dichloro-2,4,6-trifluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010448-25G 010448-5G
FLUO010448-25GEA 0 GBP
FLUO010448-25G FLUO010448-5G
3,5-Dichloro-2,4,6-trifluorobenzoic acid
3,5-Dichloro-2,4,6-trifluorobenzoic acid
Formula: C₇HCl₂F₃O₂
MDL Number: MFCD00036040
CAS Number: 13656-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR