2-Bromo-6-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1527-5G
AN-1527-1G
COBBAN-1527-5GEA
0
GBP
COBBAN-1527-5G
COBBAN-1527-1G
2-Bromo-6-nitroaniline
2-Bromo-6-nitroaniline
Formula:
C₆H₅BrN₂O₂ |
MDL Number:
MFCD09263476 CAS Number: 59255-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...