2-Bromo-5-fluorothioanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-4-fluoro-2-(methylthio)benzene

OT-0791-5G OT-0791-1G
COBBOT-0791-5GEA 379.87 GBP
COBBOT-0791-5G COBBOT-0791-1G
2-Bromo-5-fluorothioanisole
2-Bromo-5-fluorothioanisole
Formula: C₇H₆BrFS
MW: 221.09 g/mol
MDL Number: MFCD11855947
CAS Number: 147460-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR