5-Bromobenzothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2374-1G HC-2374-5G
COBBHC-2374-1GEA 0 GBP
COBBHC-2374-1G COBBHC-2374-5G
5-Bromobenzothiazole
5-Bromobenzothiazole
Formula: C₇H₄BrNS
MDL Number: MFCD03001442
CAS Number: 768-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR