1-Butylpiperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008902-1G
008902-5G
FLUO008902-1GEA
0
GBP
FLUO008902-1G
FLUO008902-5G
1-Butylpiperazine
1-Butylpiperazine
Formula:
C₈H₁₈N₂ MW: 142.24 g/mol |
MDL Number:
MFCD00039702 CAS Number: 5610-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...