2-Chloro-4-(trifluoromethyl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-1454-100G
PY-1454-1G
PY-1454-25G
PY-1454-5G
COBBPY-1454-100GEA
0
GBP
COBBPY-1454-100G
COBBPY-1454-1G
COBBPY-1454-25G
COBBPY-1454-5G
2-Chloro-4-(trifluoromethyl)pyridine
2-Chloro-4-(trifluoromethyl)pyridine
Formula:
C₆H₃ClF₃N MW: 181.54 g/mol Boiling Pt: 146…147 °C (1013 hPa) Melting Pt: –1 °C Density: 1.411 g/cm³ (20 °C) |
MDL Number:
MFCD00042224 CAS Number: 81565-18-6 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...