2-Amino-6-methylbenzothiazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2141-25G
HC-2141-100G
COBBHC-2141-25GEA
0
GBP
COBBHC-2141-25G
COBBHC-2141-100G
2-Amino-6-methylbenzothiazole
2-Amino-6-methylbenzothiazole
Formula:
C₈H₈N₂S MW: 164.23 g/mol Boiling Pt: 321 °C (1013 hPa) Melting Pt: 142 °C |
MDL Number:
MFCD00005789 CAS Number: 2536-91-6 EINECS: 219-801-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...