Molybdenum dioxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
711209-10G
711209-50G
SIAL711209-10GEA
54.6
GBP
SIAL711209-10G
SIAL711209-50G
Molybdenum dioxide, Sigma-Aldrich®
Molybdenum dioxide
Formula:
MoO₂ MW: 127.94 g/mol Boiling Pt: 1207…1307 °C (1013 hPa) Melting Pt: 770…795 °C Density: 6,47 g/cm³ (25 °C) |
MDL Number:
MFCD00014223 CAS Number: 18868-43-4 EINECS: 242-637-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...