4-Pentyn-2-ol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-Pentyn-4-ol
M-Clarity™ quality level = MQ100
268992-5G
268992-25G
SIAL268992-5GEA
75.3
GBP
SIAL268992-5G
SIAL268992-25G
4-Pentyn-2-ol, Sigma-Aldrich®
4-Pentyn-2-ol
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 126 °C (1013 hPa) Density: 0.894 g/cm³ (17 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004555 CAS Number: 2117-11-5 EINECS: 218-321-1 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...