2-Ethoxyphenol ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Hydroxy-2-ethoxybenzene, Pyrocatechol monoethyl ether, o-Ethoxyphenol

B24755.14 B24755.22 B24755.36
B24755.14EA 14.7 GBP
B24755.14 B24755.22 B24755.36
2-Ethoxyphenol ≥98%
2-Ethoxyphenol

Purity: 98% CAS No.: 94-71-3 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 86°(187°F) Odour: Phenol-like Boiling Point: 214-216° MDL No.: MFCD00002187

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 214…216 °C
Melting Pt: 24…27 °C
Density: 1.10
Flash Pt: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR