1,2-Dinitrobenzene

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

EHERC12783000EA 89.7 GBP
EHERC12783000
1,2-Dinitrobenzene
1,2-Dinitrobenzene
Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 112…115 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007093
CAS Number: 528-29-0
EINECS: 208-431-8
UN: 3443
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR