4-Iodophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008839-10G 008839-50G
FLUO008839-10GEA 0 GBP
FLUO008839-10G FLUO008839-50G
4-Iodophenol
4-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 92…93 °C
Density: 2.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002327
CAS Number: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8,III
Merck Index: 13,05058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR