1,3-Diiodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001752-10G 001752-25G
FLUO001752-10GEA 0 GBP
FLUO001752-10G FLUO001752-25G
1,3-Diiodobenzene
1,3-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2,76 g/cm³ (0 °C)
MDL Number: MFCD00041731
CAS Number: 626-00-6
EINECS: 210-921-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR