1,2-Difluoro-4-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,4-Difluoronitrobenzene

001664-25G 001664-100G 001664-500G
FLUO001664-25GEA 0 GBP
FLUO001664-25G FLUO001664-100G FLUO001664-500G
1,2-Difluoro-4-nitrobenzene
1,2-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80 °C (1013 hPa)
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007198
CAS Number: 369-34-6
EINECS: 206-718-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR