2-Fluoroaniline

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

MOLEM30255446EA 0 GBP
MOLEM30255446
2-Fluoroaniline
2-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007642
CAS Number: 348-54-9
EINECS: 206-478-9
UN: 2941
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR