2-Fluoroaniline for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

8.20565.0100EA 227 GBP
8.20565.0100 8.20565.9999
2-Fluoroaniline for synthesis, Sigma-Aldrich®
2-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007642
CAS Number: 348-54-9
EINECS: 206-478-9
UN: 2941
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 17 g/l (20 °C)
Melting Point -29 °C
Molar Mass 111.11 g/mol
Boiling Point 171 - 173 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 60 °C
Refractive Index 1.5421 (20 °C, 589 nm)
Density 1.15 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR