2,4,5-Trifluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001414-5G 001414-100G 001414-25G
FLUO001414-5GEA 0 GBP
FLUO001414-5G FLUO001414-100G FLUO001414-25G
2,4,5-Trifluoroaniline
2,4,5-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: 60 °C
MDL Number: MFCD00007649
CAS Number: 367-34-0
EINECS: 206-692-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR