2,3,4-Trifluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001805-25G 001805-100G 001805-250G
FLUO001805-25GEA 0 GBP
FLUO001805-25G FLUO001805-100G FLUO001805-250G
2,3,4-Trifluoroaniline
2,3,4-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: 14…15 °C
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00011737
CAS Number: 3862-73-5
EINECS: 407-170-9
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR