1,2-Difluoro-4-iodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,4-Difluoroiodobenzene

004819-5G 004819-25G
FLUO004819-5GEA 0 GBP
FLUO004819-5G FLUO004819-25G
1,2-Difluoro-4-iodobenzene
1,2-Difluoro-4-iodobenzene
Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 177 °C (10 mmHg)
Density: 1,99 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044630
CAS Number: 64248-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR