4-Fluoro-3-nitrobenzoic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

FLUO018022-100GEA 0 GBP
FLUO018022-100G
4-Fluoro-3-nitrobenzoic acid
4-Fluoro-3-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 92 °C (15 mmHg)
Melting Pt: 123…124 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR