2,4,6-Trifluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007485-5G 007485-100G 007485-25G
FLUO007485-5GEA 0 GBP
FLUO007485-5G FLUO007485-100G FLUO007485-25G
2,4,6-Trifluorobenzoic acid
2,4,6-Trifluorobenzoic acid
Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 144…145 °C
MDL Number: MFCD00042398
CAS Number: 28314-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR