2,3,4,5-Tetrafluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003054-25G
003054-100G
003054-500G
FLUO003054-25GEA
0
GBP
FLUO003054-25G
FLUO003054-100G
FLUO003054-500G
2,3,4,5-Tetrafluorobenzoic acid
2,3,4,5-Tetrafluorobenzoic acid
Formula:
C₇H₂F₄O₂ MW: 194.09 g/mol Boiling Pt: 135…137 °C (23 mmHg) Melting Pt: 92…92,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009613 CAS Number: 1201-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...