2,5-Difluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001935-25G 001935-100G 001935-250G
FLUO001935-25GEA 0 GBP
FLUO001935-25G FLUO001935-100G FLUO001935-250G
2,5-Difluorobenzonitrile
2,5-Difluorobenzonitrile
Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 87…92 °C (30 mmHg)
Melting Pt: 29.5…30.5 °C
MDL Number: MFCD00001777
CAS Number: 64248-64-2
EINECS: 264-753-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR