2-Fluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001671-100G
001671-250G
FLUO001671-100GEA
0
GBP
FLUO001671-100G
FLUO001671-250G
2-Fluorobenzaldehyde
2-Fluorobenzaldehyde
Formula:
C₇H₅FO MW: 124.11 g/mol Boiling Pt: 175 °C (1013 hPa) Melting Pt: –44.5 °C Density: 1.19 g/cm³ (20 °C) Flash Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003302 CAS Number: 446-52-6 EINECS: 207-171-2 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...