1,2,3,4-Tetrafluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003310-1G
003310-5G
003310-25G
FLUO003310-1GEA
0
GBP
FLUO003310-1G
FLUO003310-5G
FLUO003310-25G
1,2,3,4-Tetrafluorobenzene
1,2,3,4-Tetrafluorobenzene
Formula:
C₆H₂F₄ MW: 150.08 g/mol Boiling Pt: 95 °C (1013 hPa) Melting Pt: –42 °C Density: 1.811 g/cm³ (20 °C) |
MDL Number:
MFCD00000285 CAS Number: 551-62-2 EINECS: 208-997-6 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...