1,2,3,4-Tetrafluorobenzene ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

B21787.06 B21787.14 B21787.22
B21787.06EA 61.3 GBP
B21787.06 B21787.14 B21787.22
1,2,3,4-Tetrafluorobenzene ≥99%
1,2,3,4-Tetrafluorobenzene
Formula: C₆H₂F₄
MW: 150.08 g/mol
Boiling Pt: 94…96 °C
Melting Pt: –42 °C
Density: 1.400
Flash Pt: 20 °C (69 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000285
CAS Number: 551-62-2
EINECS: 208-997-6
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR