2,4,6-Trichloroanisole

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4,6-Trichloro-1-methoxybenzene1,3,5-Trichloro-2-methoxybenzene

EHERC17714600EA 96.8 GBP
EHERC17714600
2,4,6-Trichloroanisole
2,4,6-Trichloroanisole
Formula: C₇H₅Cl₃O
MW: 211.47 g/mol
Boiling Pt: 235…238 °C (1013 hPa)
Melting Pt: 60…61 °C
Density: 1,64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000588
CAS Number: 87-40-1
EINECS: 201-743-5
Merck Index: 13,09702

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR