5-Chloro-2-nitroanisole
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLEM46260108EA
0
GBP
MOLEM46260108
5-Chloro-2-nitroanisole
5-Chloro-2-nitroanisole
Formula:
C₇H₆ClNO₃ MW: 187.58 g/mol Melting Pt: 71 °C |
MDL Number:
MFCD00007288 CAS Number: 6627-53-8 EINECS: 229-594-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...