5-Chloro-2-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

008151-100G 008151-25G 008151-250G
FLUO008151-100GEA 0 GBP
FLUO008151-100G FLUO008151-25G FLUO008151-250G
5-Chloro-2-fluoroaniline
5-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD00069416
CAS Number: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR