2,4-Dichloro-5-fluoro-3-nitrobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023234-25G 023234-100G
FLUO023234-25GEA 0 GBP
FLUO023234-25G FLUO023234-100G
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid
Formula: C₇H₂Cl₂FNO₄
MW: 254 g/mol
MDL Number: MFCD03427137
CAS Number: 106809-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR