4-Chloro-3-fluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002198-5G
002198-25G
FLUO002198-5GEA
0
GBP
FLUO002198-5G
FLUO002198-25G
4-Chloro-3-fluorobenzaldehyde
4-Chloro-3-fluorobenzaldehyde
Formula:
C₇H₄ClFO MW: 158.56 g/mol |
MDL Number:
MFCD00143288 CAS Number: 5527-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...