4-Chloro-3-fluorobenzaldehyde ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
41365.06EA
89.6
GBP
41365.06
41365.03
41365.14
4-Chloro-3-fluorobenzaldehyde ≥98%
4-Chloro-3-fluorobenzaldehyde
Purity: 98%
CAS No.: 5527-95-7
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point: 96°(206°F)
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 46-48°
MDL No.: MFCD00143288
Formula:
C₇H₄ClFO MW: 158.56 g/mol Melting Pt: 46…48 °C Flash Pt: 96 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00143288 CAS Number: 5527-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...