3-Chloro-4-fluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002148-5G
002148-25G
FLUO002148-5GEA
0
GBP
FLUO002148-5G
FLUO002148-25G
3-Chloro-4-fluorobenzaldehyde
3-Chloro-4-fluorobenzaldehyde
Formula:
C₇H₄ClFO MW: 158.56 g/mol Boiling Pt: 66 °C (2 mmHg) Melting Pt: 44…46 °C |
MDL Number:
MFCD00011735 CAS Number: 34328-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...