1-Chloro-4-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Chloro-1-fluoro-4-benzène, 4-Chlorofluorobenzene

001147-100G 001147-500G
FLUO001147-100GEA 0 GBP
FLUO001147-100G FLUO001147-500G
1-Chloro-4-fluorobenzene
1-Chloro-4-fluorobenzene
Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –21.5 °C
Density: 1,2573 g/cm³ (0 °C)
MDL Number: MFCD00000603
CAS Number: 352-33-0
EINECS: 206-521-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR