1-Chloro-3-fluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001146-100G
001146-500G
001146-25G
FLUO001146-100GEA
0
GBP
FLUO001146-100G
FLUO001146-500G
FLUO001146-25G
1-Chloro-3-fluorobenzene
1-Chloro-3-fluorobenzene
Formula:
C₆H₄ClF MW: 130.55 g/mol Boiling Pt: 126 °C (1013 hPa) Density: 1,221 g/cm³ (25 °C) |
MDL Number:
MFCD00000569 CAS Number: 625-98-9 EINECS: 210-919-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...