5-Chloro-1,3-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-3,5-difluorobenzene

001484-5G 001484-25G
FLUO001484-5GEA 0 GBP
FLUO001484-5G FLUO001484-25G
5-Chloro-1,3-difluorobenzene
5-Chloro-1,3-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 117 °C (1013 hPa)
Density: 1,329 g/cm³ (20 °C)
MDL Number: MFCD00041518
CAS Number: 1435-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR