2-Chloro-1,4-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Difluorochlorobenzene

001596-100G 001596-25G
FLUO001596-100GEA 0 GBP
FLUO001596-100G FLUO001596-25G
2-Chloro-1,4-difluorobenzene
2-Chloro-1,4-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 128 °C (1013 hPa)
Melting Pt: –24,6 °C
Density: 1,3561 g/cm³ (20 °C)
MDL Number: MFCD00042571
CAS Number: 2367-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR