4-Chlorosalicylic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Chloro-2-hydroxybenzoic acid

C2164.10 C2164.50
XXXXC2164.10EA 0 GBP
XXXXC2164.10 XXXXC2164.50
4-Chlorosalicylic acid
4-Chlorosalicylic acid
Formula: C₇H₅ClO₃
MW: 172.57 g/mol
MDL Number: MFCD00002449
CAS Number: 5106-98-9
EINECS: 225-829-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR