4-Amino-3-chlorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

021888-5G 021888-1G
FLUO021888-5GEA 0 GBP
FLUO021888-5G FLUO021888-1G
4-Amino-3-chlorobenzoic acid
4-Amino-3-chlorobenzoic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00460423
CAS Number: 2486-71-7
EINECS: 219-630-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR