5-Amino-2-chlorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
020066-100G
020066-25G
FLUO020066-100GEA
0
GBP
FLUO020066-100G
FLUO020066-25G
5-Amino-2-chlorobenzoic acid
5-Amino-2-chlorobenzoic acid
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Boiling Pt: 330 °C (1013 hPa) Melting Pt: 188 °C Density: 1,519 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007845 CAS Number: 89-54-3 EINECS: 201-916-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...