3,4-Dichlorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018515-5G 018515-25G
FLUO018515-5GEA 0 GBP
FLUO018515-5G FLUO018515-25G
3,4-Dichlorobenzonitrile
3,4-Dichlorobenzonitrile
Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00016379
CAS Number: 6574-99-8
EINECS: 229-494-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR